Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Calpain 11 Monoclonal Antibody (2G2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA521265
Description
Peptide Sequence: LLNRMAIKFK SFKTKGFGLD ACRCMINLMD KDGSGKLGLL EFKILWKKLK KWMDIFRECD QDHSGTLNSY EMRLVIEKAG IKLNNKVMQV LVARY.
The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
Specifications
Calpain 11 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9UMQ6 | |
CAPN11 | |
CAPN11 (NP_008989, 557 a.a. ~ 651 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
2G2 | |
Unconjugated | |
CAPN11 | |
calcium-activated neutral proteinase 11; calcium-activated neutral proteinase 11 {ECO:0000250; calcium-dependent thiol protease; calpain 11; calpain 11 {ECO:0000312; calpain11; Calpain-11; calpain-11; LOW QUALITY PROTEIN: calpain-11; CANP 11; CANP 11 {ECO:0000250; Capn11; capn11 {ECO:0000312; EMBL:AAH97256.1}; HIP2; LIG; RGD:1302946}; UniProtKB:Q9UMQ6} | |
Mouse | |
Affinity Chromatography | |
RUO | |
11131 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction