Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain 11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543793
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543793 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543793 Supplier Invitrogen™ Supplier No. PA543793
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NNSRLKAKGV GQHDNAQNFG NQSFEELRAA CLRKGELFED PLFPAEPSSL Sequence homology: Cow: 85%; Dog: 100%; Goat: 85%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 77%; Rabbit: 93%; Rat: 91%; Sheep: 85%; Zebrafish: 92%.

The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CAPN11
Gene Accession No. Q9UMQ6
Gene Alias calcium-activated neutral proteinase 11; calcium-activated neutral proteinase 11 {ECO:0000250; calcium-dependent thiol protease; calpain 11; calpain 11 {ECO:0000312; calpain11; Calpain-11; calpain-11; LOW QUALITY PROTEIN: calpain-11; CANP 11; CANP 11 {ECO:0000250; Capn11; capn11 {ECO:0000312; EMBL:AAH97256.1}; HIP2; LIG; RGD:1302946}; UniProtKB:Q9UMQ6}
Gene Symbols CAPN11
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CAPN11 (aa 43-92).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11131
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.