Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain 12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562775
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562775 Supplier Invitrogen™ Supplier No. PA562775
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDEADFTLRV FSERRHTAVE IDDVISADLQ SLQGPYLPLE LGLEQLFQEL AGEEEELNAS QLQALLSIAL EPARAHTSTP REIGLRT Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 80%.

The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 12
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAPN12
Gene Accession No. Q6ZSI9
Gene Alias calcium-activated neutral proteinase 12; calpain 12; calpain-12; CANP 12; CAPN12
Gene Symbols CAPN12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Calpain 12. Recombinant protein control fragment (Product #RP-104095).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147968
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.