Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain 13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556171
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556171 Supplier Invitrogen™ Supplier No. PA556171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DIDATQLQGL LNQELLTGPP GDMFSLDECR SLVALMELKV NGRLDQEEFA RLWKRLVHYQ HVFQKVQTSP GVLLSSDLWK AIENTDFLRG IF Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 73%.

The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 13
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAPN13
Gene Accession No. Q6MZZ7
Gene Alias Calcium-activated neutral proteinase 13; calpain 13; calpain-13; CANP 13; CAPN13
Gene Symbols CAPN13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Calpain 13. Recombinant protein control fragment (Product #RP-91752).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92291
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.