Learn More
Invitrogen™ Calpain 13 Polyclonal Antibody

Description
Immunogen sequence: DGEFWMSCQDF QQKFIAMFIC SEIPITLDHG NTLHEGWSQI MFRKQVILGN TAGGPRNDAQ FNFSVQEPME GTNVVVCVTV AVTPSNLKAE DA Highest antigen sequence identity to the following orthologs - mouse 55%, rat 55%.
Specifications
Specifications
| Antigen | Calpain 13 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CAPN13 |
| Gene Accession No. | Q6MZZ7 |
| Gene Alias | Calcium-activated neutral proteinase 13; calpain 13; calpain-13; CANP 13; CAPN13 |
| Gene Symbols | CAPN13 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.