Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain 15 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232098AF488
Description
Calpain 15 Polyclonal antibody specifically detects Calpain 15 in Human samples. It is validated for Western BlotSpecifications
Calpain 15 | |
Polyclonal | |
Western Blot | |
calpain-15, CAPN15Small optic lobes homolog, EC 3.4.22, EC 3.4.22.-, EC 3.4.22.54, MGC131491, small optic lobes (Drosophila) homolog, small optic lobes homolog (Drosophila) | |
The immunogen this antibody was made to, maps to a region between residue 25 to 75 of human Small Optic Lobes Homolog using the numbering given in entry NP_005623.1 (GeneID 6650). | |
0.1 mL | |
Vision | |
6650 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction