Learn More
Invitrogen™ Calpain 15 Monoclonal Antibody (4E2)

Description
Peptide Sequence: HPKAYLHVQC DCTDSFNVVS TRGSLRTQDS VPPLHRQVLV ILSQLEGNAG FSITHRLAHR KAAQAFLSDW TASKGTHSPP LTPEVAGLHG PRPL.
Specifications
Specifications
| Antigen | Calpain 15 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4E2 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CAPN15 |
| Gene Accession No. | O75808 |
| Gene Alias | calpain 15; calpain-15; CAPN15; LOW QUALITY PROTEIN: calpain-15; small optic lobes homolog; Solh; zmp:0000001110 |
| Gene Symbols | CAPN15 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.