Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain 15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552888 Supplier Invitrogen™ Supplier No. PA552888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLPVLNGVLP KPPAILGEPK GSCQEEAGPV RTAGLVATEP ARGQCEDKDE EEKEEQEEEE GAAEPRGGWA CPRCTLHNTP VASSCSVCGG PRRLSLPRIP PEALVVPE Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.

The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 15
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAPN15
Gene Accession No. O75808
Gene Alias calpain 15; calpain-15; CAPN15; LOW QUALITY PROTEIN: calpain-15; small optic lobes homolog; Solh; zmp:0000001110
Gene Symbols CAPN15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Calpain 15. Recombinant protein control fragment (Product #RP-89312).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6650
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.