Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain 2 Antibody (17E9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Calpain 2 |
---|---|
Clone | 17E9 |
Dilution | Western Blot 1:500-1:2000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
Calpain 2 Monoclonal antibody specifically detects Calpain 2 in Human samples. It is validated for Western Blot, ELISASpecifications
Calpain 2 | |
Western Blot 1:500-1:2000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Calcium-activated neutral proteinase 2, calpain 2, (m/II) large subunit, calpain 2, large [catalytic] subunit, calpain 2, large subunit, Calpain large polypeptide L2, Calpain M-type, calpain, large polypeptide L2, calpain-2 catalytic subunit, Calpain-2 large subunit, CANP 2, CANPL2CANP2, CANPml, EC 3.4.22, EC 3.4.22.53, FLJ39928, M-calpain, mCANP, Millimolar-calpain | |
A synthesized peptide derived from Human Calpain 2 [UniProt P17655] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
17E9 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Apoptosis, Cancer, Cell Biology, Endocrinology, Immunology, Neurodegeneration, Neuroscience, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
824 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title