Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain 2 Antibody (OTI3G1), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270334AF700
Description
Calpain 2 Monoclonal specifically detects Calpain 2 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Calpain 2 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CAPN2 | |
Full length recombinant protein of human Calpain 2 (NP_001739) produced in HEK293T cells. | |
0.1 mL | |
824.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3G1 | |
Western Blot, Immunohistochemistry | |
Calcium-activated neutral proteinase 2, calpain 2, (m/II) large subunit, calpain 2, large [catalytic] subunit, calpain 2, large subunit, Calpain large polypeptide L2, Calpain M-type, calpain, large polypeptide L2, calpain-2 catalytic subunit, Calpain-2 large subunit, CANP 2, CANPL2CANP2, CANPml, EC 3.4.22, EC 3.4.22.53, FLJ39928, M-calpain, mCANP, Millimolar-calpain | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction