Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578912
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578912 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578912 Supplier Invitrogen™ Supplier No. PA578912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human A431 whole cell, human Hela whole cell, rat kidney tissue, rat brain tissue, mouse kidney tissue, mouse brain tissue. IHC: human intestinal cancer tissue, human lung cancer tissue, rat small intestine tissue. ICC/IF: HELA cell. Flow: U87 cell.

Mu-Calpain, also known as Calpain-I, and m-calpain, also known as Calpain-II, are intracellular, calcium-dependent cysteine proteases. Mu-calpain has a micromolar sensitivity (thus the mu-) as compared to the millimolar calcium sensitivity of m-calpain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene CAPN2
Gene Accession No. O08529, P17655, Q07009
Gene Alias 80 kDa M-calpain subunit; AI326419; Calcium-activated neutral proteinase 2; calcium-dependent protease; CALP80; calpain 2; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; calpain II; calpain large polypeptide L2; Calpain M-type; calpain, large polypeptide L2; calpain-2 catalytic subunit; Calpain-2 large subunit; CANP 2; CANP2; CANPL2; CANPml; Capa2; Capa-2; CAPN2; cysteine proteinase; m-calpain; m-calpain 80 kDa large subunit; m-calpain large subunit; m-calpin; mCANP; millimolar-calpain
Gene Symbols CAPN2
Host Species Rabbit
Immunogen E. coli-derived human Calpain 2 recombinant protein (Position: R500-L700).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12334, 29154, 824
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.