Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595965 Supplier Invitrogen™ Supplier No. PA595965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDTYKKWKLT KMDGNWRRGS TAGGCRNYPN TFWMNPQYLI KLEEEDEDEE DGESGCTFLV GLIQKHRRRQ RKMGEDMHTI GFGIYEVPEE LSGQTNIHLS KNFFLTNRAR ERSDTFINLR EVLNRFKLPP GEYILVPSTF EPNKDGDFCI RVFSEKKADY QAVDDEIEAN LEEFDISEDD IDDGFRRLFA QLAGEDAEIS AFELQTILRR VLAKRQDIKS DGFSIETCKI MVDMLDSDGS GKLGLKEFYI LWTKIQKYQK IYREIDVDRS GTMNSYEMRK ALEEAGFKMP CQLHQVIVAR FADDQLIIDF DNFVRCLVRL ETLFKIFKQL D; Positive Samples: H460, A-549, HeLa, Mouse kidney, Mouse lung; Cellular Location: Cell membrane, Cytoplasm.

Mu-Calpain, also known as Calpain-I, and m-calpain, also known as Calpain-II, are intracellular, calcium-dependent cysteine proteases. Mu-calpain has a micromolar sensitivity (thus the mu-) as compared to the millimolar calcium sensitivity of m-calpain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CAPN2
Gene Accession No. O08529, P17655, Q07009
Gene Alias 80 kDa M-calpain subunit; AI326419; Calcium-activated neutral proteinase 2; calcium-dependent protease; CALP80; calpain 2; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; calpain II; calpain large polypeptide L2; Calpain M-type; calpain, large polypeptide L2; calpain-2 catalytic subunit; Calpain-2 large subunit; CANP 2; CANP2; CANPL2; CANPml; Capa2; Capa-2; CAPN2; cysteine proteinase; m-calpain; m-calpain 80 kDa large subunit; m-calpain large subunit; m-calpin; mCANP; millimolar-calpain
Gene Symbols CAPN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-680 of human Calpain 2 (NP_0017392).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12334, 29154, 824
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.