Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 104151AP150UL

Description
Calpain, a heterodimer consisting of a large and a small subunit, is a major intracellular protease, although its function has not been well established. This gene encodes a muscle-specific member of the calpain large subunit family that specifically binds to titin. Mutations in this gene are associated with limb-girdle muscular dystrophies type 2A. Alternate promoters and alternative splicing result in multiple transcript variants encoding different isoforms and some variants are ubiquitously expressed.Specifications
Calpain 3 | |
Polyclonal | |
Unconjugated | |
CAPN3 | |
Calpain 3, calpain 3, (p94), Calpain L3, Calpain p94, CANP 3, CANP3, CANPL3, CAPN3, LGMD2, LGMD2A, nCL 1, NCL1, New calpain 1, p94 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12335, 29155, 825 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P16259, P20807, Q64691 | |
Capn3 | |
Calpain 3 Fusion Protein Ag0671 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction