Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain 5 Monoclonal Antibody (OTI7D2)
GREENER_CHOICE

Catalog No. PIMA526693
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526693 Supplier Invitrogen™ Supplier No. MA526693
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calpain 5 Monoclonal Antibody for IHC (P)

Calpains are calcium-dependent cysteine proteases involved in signal transduction in a variety of cellular processes. A functional calpain protein consists of an invariant small subunit and 1 of a family of large subunits. CAPN5 is one of the large subunits. Unlike some of the calpains, CAPN5 and CAPN6 lack a calmodulin-like domain IV. Because of the significant similarity to Caenorhabditis elegans sex determination gene tra-3, CAPN5 is also called as HTRA3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 5
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI7D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CAPN5
Gene Accession No. O15484
Gene Alias ADNIV; calpain 5; calpain htra-3; Calpain-5; Capn5; high-temperature requirement factor A3; HTRA3; Ncl3; nCL-3; New calpain 3; testis tissue sperm-binding protein Li 91mP; VRNI
Gene Symbols CAPN5
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 274-578 of CAPN5 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 726
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.