Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain 5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121098
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121098 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121098 Supplier Invitrogen™ Supplier No. PA5121098
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SKOV3, Mouse thymus, Mouse brain, Mouse spinal cord. Immunogen sequence: GAWTLHEDPR QNRGGGCINH KDTFFQNPQY IFEVKKPEDE VLICIQQRPK RSTRREGKGE NLAIGFDIYK VEENRQYRMH SLQHKAASSI YINSRSVFLR TDQPEGRYVI IPTTFEPGHT GEFLLRVFTD VPSNCRELRL DEPPHTCWSS LCGYPQLVTQ VHVLGAAGLK DSPTGANSYV IIKCEGDKVR SAVQKGTSTP EYNVKGIFYR KKLSQPITVQ VWNHRVLKDE FLGQVHLKAD PDNLQALHTL HLRDRNSRQP SNLPGTVAVH ILSSTSLMAV.

Calpains are calcium-dependent cysteine proteases involved in signal transduction in a variety of cellular processes. A functional calpain protein consists of an invariant small subunit and 1 of a family of large subunits. CAPN5 is one of the large subunits. Unlike some of the calpains, CAPN5 and CAPN6 lack a calmodulin-like domain IV. Because of the significant similarity to Caenorhabditis elegans sex determination gene tra-3, CAPN5 is also called as HTRA3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CAPN5
Gene Accession No. O08688, O15484, Q8R4C0
Gene Alias ADNIV; calpain 5; calpain htra-3; Calpain-5; Capn5; high-temperature requirement factor A3; HTRA3; Ncl3; nCL-3; New calpain 3; testis tissue sperm-binding protein Li 91mP; VRNI
Gene Symbols CAPN5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 361-640 of human CAPN5 (NP_004046.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12337, 171495, 726
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.