Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain 5 Recombinant Rabbit Monoclonal Antibody (HL2432), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Thermo Scientific MA550530

Description
Calpains are calcium-dependent cysteine proteases involved in signal transduction in a variety of cellular processes. A functional calpain protein consists of an invariant small subunit and 1 of a family of large subunits. CAPN5 is one of the large subunits. Unlike some of the calpains, CAPN5 and CAPN6 lack a calmodulin-like domain IV. Because of the significant similarity to Caenorhabditis elegans sex determination gene tra-3, CAPN5 is also called as HTRA3.
Specifications
Calpain 5 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
O08688, O15484 | |
CAPN5 | |
Recombinant fragment of human Calpain 5. | |
100 μL | |
Primary | |
Human, Mouse | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
HL2432 | |
Unconjugated | |
CAPN5 | |
ADNIV, calpain 5, calpain htra-3, Calpain-5, Capn5, high-temperature requirement factor A3, HTRA3, Ncl3, nCL-3, New calpain 3, testis tissue sperm-binding protein Li 91mP, VRNI | |
Rabbit | |
Protein A | |
RUO | |
12337, 726 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction