Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain 6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583496
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583496 Supplier Invitrogen™ Supplier No. PA583496
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLDMPKMSCW NLARGYPKVV TQITVHSAED LEKKYANETV NPYLVIKCGK EEVRSPVQKN TVHAIFDTQA IFYRRTTDIP IIVQVWNSRK FCDQFLGQ Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.

Calpain-6, also known as calpamodulin or calpain-X, is an intracellular, calcium-dependent cysteine protease. Calpain-6 has a much more tissue-specific expression in adults than the ubiquitous calpain-1 and calpain-2, and has thus far been found primarily in the placenta, although it is expressed embryonically in a number of tissues. The classical calpain family members consist of a common small subunit (calpain-4), and a large variable subunit, but it is not known if calpain-6 associates with a small subunit. Domains in the large subunit include the amino terminal domain-I, the proteinase domain-II, domain-III, and the EF-hand domain-IV (domain T in calpains 5 and 6). Located on the X chromosome, the calpain-6 sequence lacks the 'EF hand' calcium-binding motif found in domain-IV of the classical calpains. In addition, the canonical active site Cys, His and Asn are modified to Lys, His, Asn in human and Lys, Tyr, Asn in mouse calpain-6, making it unlikely that calpain-6 is proteolytically active.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAPN6
Gene Accession No. Q9Y6Q1
Gene Alias calpain 6; calpain-6; Calpain-like protease X-linked; calpamodulin; CalpM; CANPX; Capa6; CAPN6; CAPNX; DJ914P14.1
Gene Symbols CAPN6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CAPN6. Recombinant protein control fragment (Product #RP-98332).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 827
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.