Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain 7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561260
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561260 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561260 Supplier Invitrogen™ Supplier No. PA561260
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVFYYKEAAQ ALIYAEMAGS SLENIQEKIT EYLERVQALH SAVQSKSADP LKSKHQLDLE RAHFLVTQAF DEDEKENVED AIELYTEAVD LCLKTSYETA DKVLQNK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.

Calpain proteinases are defined currently as papain-like neutral proteases that are calcium activated. Calpains 1 and 2 are composed of a large subunit, which is proteolytically active, and a small subunit (also called calpain-4), or enhancer protein, that is not proteolytically active. The other calpain proteins identified to date (calpain 5, 6, 8, 9, 10, 11, 12, 13, 14, 15) are not known to require a small subunit. Domains in the large subunit include the amino terminal domain-I, the proteinase domain-II, domain-III, and the EF-hand domain-IV (domain-T in calpains 5 & 6, domain-N for calpain-7). Calpain-7 domain-N does not have the standard calpain EF hand domain for binding calcium, making it unclear whether it is calcium regulated. In place of the EF hand calcium-binding domain IV, calpain-7 has a sequence most closely resembling domain-III, with unknown function. Calpain-7 is widely distributed, found in most tissues analyzed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAPN7
Gene Accession No. Q9Y6W3
Gene Alias AU022319; calpain 7; calpain like protease; CALPAIN7; calpain-7; Capn7; homolog of Aspergillus Nidulans PALB; palB homolog; PALBH
Gene Symbols CAPN7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Calpain 7. Recombinant protein control fragment (Product #RP-88629).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23473
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.