Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain 7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116762 Supplier Thermo Scientific Supplier No. PA5116762
Add to Cart
Add to Cart

Calpain 7 Polyclonal Antibody

Calpain 7 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 7
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CAPN7
Gene Accession No. Q9R1S8, Q9Y6W3
Gene Alias AU022319; calpain 7; calpain like protease; CALPAIN7; calpain-7; Capn7; homolog of Aspergillus Nidulans PALB; palB homolog; PALBH
Gene Symbols CAPN7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-220 of human CAPN7 (NP_0551111).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12339, 23473, 306260
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.