Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain 9 Antibody (OTI3G10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271626H
Description
Calpain 9 Monoclonal specifically detects Calpain 9 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Calpain 9 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CAPN9 | |
Full length recombinant protein of human Calpain 9 (NP_006606) produced in HEK293T cells. | |
0.1 mL | |
10753.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3G10 | |
Western Blot, Immunohistochemistry | |
calpain 9, calpain 9 (nCL-4), calpain-9, Digestive tract-specific calpain, EC 3.4.22, EC 3.4.22.54, GC36, NCL4, nCL-4EC 3.4.22.-, New calpain 4, novel calpain large subunit-4, Protein CG36 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction