Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain 9 Antibody (OTI3G10), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271626PCP
Description
Calpain 9 Monoclonal antibody specifically detects Calpain 9 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Calpain 9 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Western Blot, Immunohistochemistry | |
OTI3G10 | |
Western Blot, Immunohistochemistry | |
calpain 9, calpain 9 (nCL-4), calpain-9, Digestive tract-specific calpain, EC 3.4.22, EC 3.4.22.54, GC36, NCL4, nCL-4EC 3.4.22.-, New calpain 4, novel calpain large subunit-4, Protein CG36 | |
Full length human recombinant protein of human CAPN9(NP_006606) produced in HEK293T cell. | |
0.1 mL | |
Cell Biology, Neuroscience, Signal Transduction | |
10753 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction