Learn More
Invitrogen™ Calpain 9 Monoclonal Antibody (OTI3G10)

Description
Calpain 9 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | Calpain 9 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3G10 |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CAPN9 |
| Gene Accession No. | O14815, O35920 |
| Gene Alias | 2200003B16Rik; calpain 9; calpain-9; Capn9; Digestive tract-specific calpain; GC36; NCL4; nCL-4; New calpain 4; novel calpain large subunit-4; Protein CG36 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.