Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain Rabbit anti-Flatworm, Polyclonal, FabGennix

Catalog No. 502206336
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6336 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6336 Supplier Fabgennix Inc Supplier No. CALP101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calpain Polyclonal antibody specifically detects Calpain in Flatworm samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen Calpain
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Alias calcium activated neutral protease; calcium-activated neutral protease 1; calcium-activated neutral proteinase 1; calcium-dependent protease; calpain; calpain 1; calpain 1 large subunit; calpain 1, (mu/I) large subunit; calpain 1, large subunit; calpain mu-type; calpain, large polypeptide L1; calpain-1 catalytic subunit; Calpain-1 large subunit; Calpain-I; Calpain-II; CANP; CANP 1; Canp1; CANPL1; Capa1; Capa-1; Capn1; Cell proliferation-inducing gene 30 protein; cell proliferation-inducing protein 30; Cls1; I79_024883; micromolar calcium-activated neutral protease 1; micromolar calcium-activated neutral protease 1 isoform A; micromolar calcium-activated neutral protease 1 isoform B; micromolar calcium-dependent neutral protease large subunit; micromolar-calpain; mucalpain; mu-calpain; mu-calpin; muCANP; muCL; PIG30; SPG76
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 700-758 on Schistosoma mansoni Calpain protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Flatworm
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.