Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain S1 Antibody (1D11), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259446FR
This item is not returnable.
View return policy
Description
Calpain S1 Monoclonal antibody specifically detects Calpain S1 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Calpain S1 | |
Monoclonal | |
DyLight 680 | |
Calcium-activated neutral proteinase small subunit, Calcium-dependent protease small subunit 1, calcium-dependent protease, small subunit, calpain 4, small subunit (30K), Calpain regulatory subunit, calpain small subunit 1, calpain, small polypeptide, CALPAIN4, CANP, CANPS, CAPN4, CDPS, CSS1, epididymis secretory sperm binding protein | |
Recombinant human Calpain S1 (84-268aa) purified from E. coli | |
0.1 mL | |
Cell Cycle and Replication | |
826 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunofluorescence | |
1D11 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction