Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain S1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117054
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117054 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117054 Supplier Thermo Scientific Supplier No. PA5117054
Add to Cart
Add to Cart

Calpain S1 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain S1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CAPNS1
Gene Accession No. O88456, P04632, Q64537
Gene Alias 30K; Calcium-activated neutral proteinase small subunit; calcium-dependent protease; Calcium-dependent protease small subunit; Calcium-dependent protease small subunit 1; calcium-dependent protease, small subunit; calpain 4; calpain 4, small subunit (30K); calpain II regulatory subunit; calpain II regulatory subunit (EC 3.4.22.17); calpain regulatory subunit; Calpain small subunit 1; calpain, small polypeptide; calpain, small subunit 1; CALPAIN4; CANP; CANP small subunit; CANPS; Capa4; Capa-4; Capn4; CAPNS; CAPNS1; CDPS; CSS1; D7Ertd146e; small pol; unknown protein
Gene Symbols CAPNS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 69-268 of human Calpain small subunit 1 (NP_0017401).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12336, 29156, 826
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.