Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain S1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531110 Supplier Invitrogen™ Supplier No. PA531110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T. Predicted reactivity: Mouse (89%), Pig (97%), Rabbit (96%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Calpains are a ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. Calpain families have been implicated in neurodegenerative processes, as their activation can be triggered by calcium influx and oxidative stress. Calpain I and II are heterodimeric with distinct large subunits associated with common small subunits, all of which are encoded by different genes. Calpain S1 is a small subunit common to both calpain I and II and is associated with myotonic dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain S1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene CAPNS1
Gene Accession No. P04632
Gene Alias 30K; Calcium-activated neutral proteinase small subunit; calcium-dependent protease; Calcium-dependent protease small subunit; Calcium-dependent protease small subunit 1; calcium-dependent protease, small subunit; calpain 4; calpain 4, small subunit (30K); calpain II regulatory subunit; calpain II regulatory subunit (EC 3.4.22.17); calpain regulatory subunit; Calpain small subunit 1; calpain, small polypeptide; calpain, small subunit 1; CALPAIN4; CANP; CANP small subunit; CANPS; Capa4; Capa-4; Capn4; CAPNS; CAPNS1; CDPS; CSS1; D7Ertd146e; small pol; unknown protein
Gene Symbols CAPNS1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Calpain S1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 826
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.