Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpastatin Antibody (OTI1E7), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270338MFV610
Description
Calpastatin Monoclonal antibody specifically detects Calpastatin in Human samples. It is validated for Flow CytometrySpecifications
Calpastatin | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI1E7 | |
Flow Cytometry | |
BS-17, Calpain inhibitor, calpastatin, MGC9402, Sperm BS-17 component | |
Full length human recombinant protein of human CAST(NP_001741) produced in HEK293T cell. | |
0.1 mL | |
Hypoxia, Neuroscience | |
831 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction