Learn More
Invitrogen™ Calpastatin Monoclonal Antibody (2G11D6)

Description
MA3-945 detects calpastatin from bovine, human, canine, pig, rabbit, and rat tissues and cells. This antibody does not cross-react with calpains or calmodulin. MA3-945 has been successfully used in Western blot, immunohistochemistry, and immunocytochemical procedures. By Western blot, this antibody detects 150, 125, 90 and 70 kDa proteins which have been suggested to be proteins translated from different start sites of the calpastatin gene. Immunofluorescence staining of calpastatin in porcine LLC-PK1 cells with MA3-945 results in diffuse cytoplasmic staining and weak nuclear staining. The MA3-945 antigen is bovine skeletal muscle calpastatin. This antibody recognizes an epitope found between amino acid residues 295-501 (domain II) of human calpastatin.
Specifications
Specifications
| Antigen | Calpastatin |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2G11D6 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites, PBS with 0.05% sodium azide |
| Gene | Cast |
| Gene Accession No. | P08855, P12675, P20810, P20811, P27321 |
| Gene Alias | BS-17; Calpain inhibitor; calpastatin; calpastatin isoform III; calpastatin precursor (EC 3.4.22.17); calpastatin type 1; calpastatin type 2; calpastatin type 3; calpastatin type I; calpastatin type II; calpastatin type III; calpastatin type IV; Cast; Erythrocyte calpastatin; heart calpastatin; PLACK; Sperm BS-17 component; type III calpastatin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.