Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpastatin Monoclonal Antibody (A6B1)
GREENER_CHOICE

Catalog No. PIMA544975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544975 Supplier Invitrogen™ Supplier No. MA544975
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calpastatin Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Calpastatin is an endogenous calpain (calcium-dependent cysteine protease) inhibitor. It consists of an N-terminal domain L and four repetitive calpain-inhibition domains (domains 1-4), and it is involved in the proteolysis of amyloid precursor protein. The calpain/calpastatin system is involved in numerous membrane fusion events, such as neural vesicle exocytosis and platelet and red-cell aggregation. The protein is also thought to affect the expression levels of genes encoding structural or regulatory proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpastatin
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone A6B1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Cast
Gene Accession No. P20810
Gene Alias BS-17; Calpain inhibitor; calpastatin; calpastatin isoform III; calpastatin precursor (EC 3.4.22.17); calpastatin type 1; calpastatin type 2; calpastatin type 3; calpastatin type I; calpastatin type II; calpastatin type III; calpastatin type IV; Cast; Erythrocyte calpastatin; heart calpastatin; PLACK; Sperm BS-17 component; type III calpastatin
Gene Symbols Cast
Host Species Mouse
Immunogen Recombinant protein within human Calpastatin aa 501-708/708.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 831
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.