Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpastatin Mouse anti-Human, Clone: CAST/1550, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen Calpastatin
Clone CAST/1550
Concentration 0.2 mg/ml
Dilution ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122268
SDP
View Documents
Novus Biologicals
NBP30739720UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

Calpastatin Monoclonal specifically detects Calpastatin in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.
Specifications

Specifications

Calpastatin
0.2 mg/ml
ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Hypoxia, Neuroscience
10 mM PBS with 0.05% BSA
831
IgG1 κ
Protein A or G purified
CAST/1550
ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
BS-17, Calpain inhibitor, calpastatin, MGC9402, Sperm BS-17 component
Recombinant full-length human Calpastatin protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.