Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpastatin Mouse anti-Human, Clone: CAST/1550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP307397100UG
Description
Calpastatin Monoclonal specifically detects Calpastatin in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
Calpastatin | |
Monoclonal | |
0.2 mg/mL | |
ELISA 2 to 4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
BS-17, Calpain inhibitor, calpastatin, MGC9402, Sperm BS-17 component | |
Recombinant full-length human Calpastatin protein | |
100 μg | |
Hypoxia, Neuroscience | |
831 | |
Store at 4°C. | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin) | |
CAST/1550 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction