Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calphostin C, 98%, Thermo Scientific™

Catalog No. AC328180001
Click to view available options
Quantity:
0.1 mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121263-19-2
Molecular Formula C44H38O14
Molecular Weight (g/mol) 790.774
MDL Number MFCD00133155
InChI Key LSUTUUOITDQYNO-UHFFFAOYSA-N
Synonym calphostin c, calphostin c, cladosporium cladosporioides, 1-4,9-dihydroxy-12-2-4-hydroxyphenoxycarbonyl oxy propyl-2,6,7,11-tetramethoxy-3,10-dioxoperylen-1-yl propan-2-yl benzoate, carbonic acid, 1r-2-12-2r-2-benzoyloxy propyl-3,10-dihydro-4,9-dihydroxy-2,6,7,11-tetramethoxy-3,10-dioxo-1-perylenyl-1-methylethyl4-hydroxyphenyl ester, stereoisomer, carbonic acid, 2-12-2-benzoyloxy propyl-3,10-dihydro-4,9-dihydroxy-2,6,7,11-tetramethoxy-3,10-dioxo-1-perylenyl-1-methylethyl 4-hydroxyphenyl ester, acmc-20d0wl, 1-3,10-dihydroxy-12-2-4-hydroxyphenoxy carbonyl oxy propyl-2,6,7,11-tetramethoxy-4,9-dioxo-4,9-dihydroperylen-1-yl propan-2-yl benzoate, carbonic acid, 1r-2-12-2r-2-benzoyloxy propyl-3,10-dihydro-4,9-dihydroxy-2,6,7,11-tetramethoxy-3,10-dioxo-1-perylenyl-1-methylethyl 4-hydroxyphenyl ester, stereoisomer, calphostin c from cladosporium cladosporioides hplc , powder, 1r-2-12-2r-2-benzoyloxy propyl-3,10-dihydro-4,9-dihydroxy-2,6,7,11-tetramethoxy-3,10-dioxo-1-perylenyl-1-methylethylcarbonic acid 4-hydroxyphenyl ester
PubChem CID 2533
IUPAC Name 1-[3,10-dihydroxy-12-[2-(4-hydroxyphenoxy)carbonyloxypropyl]-2,6,7,11-tetramethoxy-4,9-dioxoperylen-1-yl]propan-2-yl benzoate
SMILES CC(CC1=C(C(=C2C(=O)C=C(C3=C4C(=CC(=O)C5=C(C(=C(C(=C45)C1=C32)CC(C)OC(=O)OC6=CC=C(C=C6)O)OC)O)OC)OC)O)OC)OC(=O)C7=CC=CC=C7

Specifications

Color Red
Assay Percent Range 98%
Quantity 0.1 mg
Formula Weight 790.8
Percent Purity ≥97.5%
Physical Form Solid
Chemical Name or Material Calphostin C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.