Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calponin 1 Antibody (2H1), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Calponin 1 |
---|---|
Clone | 2H1 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Calponin 1 Monoclonal antibody specifically detects Calponin 1 in Human, Mouse samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
Calponin 1 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, smooth muscle, calponins, basic, Sm-Calp, SMCCcalponin-1 | |
A synthesized peptide derived from Human Calponin 1 [UniProt P51911] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
2H1 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Cell Biology, Cellular Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1264 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title