Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calponin 1 Antibody (CNN1/1408R), Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50 - $560.50
Specifications
Antigen | Calponin 1 |
---|---|
Clone | CNN1/1408R |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25323320
![]() |
Novus Biologicals
NBP25323320UG |
20 μg |
Each for $304.50
|
|
|||||
NB253233100
![]() |
Novus Biologicals
NBP253233100UG |
100 μg |
Each for $560.50
|
|
|||||
NB253233200
![]() |
Novus Biologicals
NBP253233200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Calponin 1 Monoclonal specifically detects Calponin 1 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Calponin 1 | |
0.2 mg/mL | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Cytoskeleton Markers | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
1264 | |
Recombinant human full-length CNN1 protein | |
Primary | |
Store at 4C. |
CNN1/1408R | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, smooth muscle, calponins, basic, Sm-Calp, SMCCcalponin-1 | |
CNN1 | |
IgG κ | |
Protein A or G purified | |
34 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title