Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calponin 1 Antibody (CNN1/4227R), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308462AF350
Description
Calponin 1 Monoclonal antibody specifically detects Calponin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Calponin 1 | |
Monoclonal | |
Alexa Fluor 350 | |
Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, smooth muscle, calponins, basic, Sm-Calp, SMCCcalponin-1 | |
Synthetic peptide corresponding to residues on the C-terminus of human Calponin 1 was used as an immunogen. (Uniprot: P51911) | |
100 μg | |
Cancer, Cell Biology, Cellular Markers | |
1264 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
CNN1/4227R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction