Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calponin 1 Antibody (CNN1/832), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247757MFV610
Description
Calponin 1 Monoclonal antibody specifically detects Calponin 1 in Human, Rat samples. It is validated for Flow CytometrySpecifications
Calponin 1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Flow Cytometry | |
CNN1/832 | |
Flow Cytometry | |
Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, smooth muscle, calponins, basic, Sm-Calp, SMCCcalponin-1 | |
Recombinant full-length human Calponin 1 protein (Uniprot: P51911) | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers | |
1264 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction