Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calponin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586024 Supplier Invitrogen™ Supplier No. PA586024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calponin 1 Polyclonal Antibody for ICC/IF, IHC, IHC (P), Flow

Calponin, a thin filament associated protein is implicated in the regulation and modulation of smooth muscle contraction. It is capable of binding to actin, calmodulin, troponin C and tropomyosin. Calponin is expressed in smooth muscle and tissues containing significant amounts of smooth muscle. Two isoforms of calponin exist whose molecular weights are 34kDa and 29kDa. Expression of the 29kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions. It stains smooth muscle, myoepithelial cells, myofibroblasts, keratinocytes and nerve fibers. It identifies myoepithelial cells in breast lesions, and helps differentiate breast collagenous spherulosis (positive) from adenoid cystic carcinoma. Adenoid cystic carcinoma in salivary gland tumors is calponin positive.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calponin 1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene CNN1
Gene Accession No. P51911, Q08091, Q08290
Gene Alias Basic calponin; calponin; calponin 1; calponin 1, basic, smooth muscle; calponin 1, smooth muscle; calponin H1, smooth muscle; Calponin-1; calponins, basic; CN; CNN1; CnnI; epididymis secretory protein Li 14; h1-calponin; HEL-S-14; similar to calponin 1, basic, smooth muscle; Sm-Calp; SMCC; smooth muscle calponin h1
Gene Symbols CNN1
Host Species Rabbit
Immunogen Synthetic peptide within Human Calponin-1 aa 248-297.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1264, 12797, 65204
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.