Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calponin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543135 Supplier Invitrogen™ Supplier No. PA543135
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MSSAHFNRGP AYGLSAEVKN KLAQKYDHQR EQELREWIEG VTGRRIGNNF Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 93%; Rat: 100%; Sheep: 100%; Zebrafish: 77%.

Calponin, a thin filament associated protein is implicated in the regulation and modulation of smooth muscle contraction. It is capable of binding to actin, calmodulin, troponin C and tropomyosin. Calponin is expressed in smooth muscle and tissues containing significant amounts of smooth muscle. Two isoforms of calponin exist whose molecular weights are 34kDa and 29kDa. Expression of the 29kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions. It stains smooth muscle, myoepithelial cells, myofibroblasts, keratinocytes and nerve fibers. It identifies myoepithelial cells in breast lesions, and helps differentiate breast collagenous spherulosis (positive) from adenoid cystic carcinoma. Adenoid cystic carcinoma in salivary gland tumors is calponin positive.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calponin 1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CNN1
Gene Accession No. P51911, Q08091
Gene Alias Basic calponin; calponin; calponin 1; calponin 1, basic, smooth muscle; calponin 1, smooth muscle; calponin H1, smooth muscle; Calponin-1; calponins, basic; CN; CNN1; CnnI; epididymis secretory protein Li 14; h1-calponin; HEL-S-14; similar to calponin 1, basic, smooth muscle; Sm-Calp; SMCC; smooth muscle calponin h1
Gene Symbols CNN1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CNN1 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1264, 12797
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.