Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calponin 2 Antibody (OTI2B5), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270421PE
Description
Calponin 2 Monoclonal antibody specifically detects Calponin 2 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
Calponin 2 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Monkey | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
OTI2B5 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
calponin 2, Calponin H2, smooth muscle, calponin-2, Neutral calponin | |
Full length human recombinant protein of human CNN2(NP_004359) produced in HEK293T cell. | |
0.1 mL | |
Signal Transduction | |
1265 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction