Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Calponin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549995
Description
The antibody detects endogenous levels of total Calponin 2 protein.
The protein encoded by this gene, which can bind actin, calmodulin, troponin C, and tropomyosin, may function in the structural organization of actin filaments. The encoded protein could play a role in smooth muscle contraction and cell adhesion. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Calponin 2 | |
Polyclonal | |
Unconjugated | |
Cnn2 | |
AA408047; AI324678; Calpo2; calponin 2; calponin H2, smooth muscle; Calponin-2; CLP2; CNN2; h2-calponin; LOW QUALITY PROTEIN: calponin-2; neutral calponin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1265, 12798 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q08093, Q99439 | |
Cnn2 | |
Synthesized peptide derived from internal of human CNN2. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction