Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calponin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519170
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519170 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519170 Supplier Invitrogen™ Supplier No. PA519170
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

This gene encodes a protein with a markedly acidic C terminus; the basic N-terminus is highly homologous to the N-terminus of a related gene, CNN1. Members of the CNN gene family all contain similar tandemly repeated motifs. This encoded protein is associated with the cytoskeleton but is not involved in contraction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calponin 3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CNN3
Gene Accession No. Q15417
Gene Alias 1600014M03Rik; acidic calponin h3; C85854; Calpo3; calponin 3; calponin 3, acidic; calponin, acidic isoform; Calponin, non-muscle isoform; calponin-3; Cnn3; dJ639P13.2.2 (acidic calponin 3)
Gene Symbols CNN3
Host Species Goat
Immunogen Synthetic peptide sequence (HGEYQDDYPRDYQYS) corresponding to the C-terminus amino acids of CNN3 (aa 309-324).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1266
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.