Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calponin (CNN1) Monoclonal Antibody (OTI3H2), TrueMAB™, OriGene
SDP

Catalog No. 502543591
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3591 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3591 Supplier OriGene Supplier No. CF813853
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calponin (CNN1) Monoclonal antibody specifically detects Calponin (CNN1) in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Calponin (CNN1)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI3H2
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Tnfrsf17
Gene Accession No. Q02223
Gene Alias Basic calponin; calponin; calponin 1; calponin 1, basic, smooth muscle; calponin 1, smooth muscle; calponin H1, smooth muscle; Calponin-1; calponins, basic; CN; CNN1; CnnI; epididymis secretory protein Li 14; h1-calponin; HEL-S-14; similar to calponin 1, basic, smooth muscle; Sm-Calp; SMCC; smooth muscle calponin h1
Host Species Mouse
Immunogen Recombinant full length protein of human TNFRSF17/BCMA (NP_001183) produced in HEK293 cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 608
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.