Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calponin Mouse anti-Bovine, Human, Porcine, Unconjugated, Clone: CALP or N3/16, MilliporeSigma™

Catalog No. MABT1504100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT1504-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT1504-100 Supplier MilliporeSigma Supplier No. MABT1504100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Calponin, clone CALP , Cat. No. MABT1504, is a mouse monoclonal antibody that detects Calponin-1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calponin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CALP or N3/16
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Basic calponin;Calponin H1 smooth muscle
Gene Symbols CNN1
Host Species Mouse
Immunogen Crude human uterine muscle extract.
Molecular Weight of Antigen ∽36 kDa observed; 33.17 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001290
Test Specificity Clone CALP specifically detects human Calponin-1.
Target Species Bovine, Human, Pig
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.