Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CALR Rabbit anti-Human, Polyclonal, Abnova™

Catalog No. 89337958 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-958 Supplier Abnova Corporation Supplier No. PAB31679
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody.

CALR Polyclonal antibody specifically detects CALR in Human samples. It is validated for WB-Ce.

Specifications

Antigen CALR
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description calreticulin
Gene CALR
Gene Alias CRT, FLJ26680, RO, SSA, cC1qR
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human CALR
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 811
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.