Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calreticulin-2/CALR3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23339025UL
Description
Calreticulin-2/CALR3 Polyclonal specifically detects Calreticulin-2/CALR3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Calreticulin-2/CALR3 | |
Polyclonal | |
Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Q96L12 | |
CALR3 | |
This antibody was developed against a recombinant protein corresponding to amino acids: FLITDDEEYADNFGKATWGETKGPEREMDAIQAKEEMKKAREEEEEELLSGKINRHEHYFNQFHRRN | |
25 μL | |
Cancer | |
125972 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
calreticulin 2, calreticulin 3, Calreticulin-2, calreticulin-3, cancer/testis antigen 93, CRT2calreticulin-2, CT93, FLJ25355, MGC26577 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction