Learn More
Invitrogen™ Calreticulin 3 Monoclonal Antibody (OTI1E1)

Description
Calreticulin 3 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | Calreticulin 3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI1E1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Calr3 |
| Gene Accession No. | Q96L12, Q9D9Q6 |
| Gene Alias | 1700031L01Rik; 6330586I20Rik; CALR3; calreticulin 3; calreticulin-2; Calreticulin-3; calspeculin; calsperin; cancer/testis antigen 93; CMH19; CRT2; cspn; CT93; EGK_10275; testis secretory sperm-binding protein Li 226n |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.