Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calreticulin Antibody (CR213-2AG), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311672IR
Description
Calreticulin Monoclonal specifically detects Calreticulin in Human samples. It is validated for Western Blot, ELISA.Specifications
Calreticulin | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
CR213-2AG | |
Western Blot, ELISA | |
autoantigen Ro, CALR, calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA | |
Recombinant human calreticulin. | |
0.1 mL | |
Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction | |
811 | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction