Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calreticulin Monoclonal Antibody (6C6)
GREENER_CHOICE

Catalog No. PIMA547459
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547459 100 μL
PIMA547360 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA547459 Supplier Invitrogen™ Supplier No. MA547459
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calreticulin Monoclonal Antibody for Western Blot, IHC, ICC/IF

Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calreticulin
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 6C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene CALR
Gene Accession No. P14211, P18418, P27797
Gene Alias Autoantigen RO; CABP3; CALBP; calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; calreticulin precursor; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99n; I79_008346; RO; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin); SSA
Gene Symbols CALR
Host Species Mouse
Immunogen Synthetic peptides VESGSLEDDWDFLPPKKI corresponding to amino acids 191-208 of human calreticulin, including the LC3 interacting region or LIR.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12317, 64202, 811
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.