Learn More
Invitrogen™ Calreticulin Recombinant Rabbit Monoclonal Antibody (SAIC-16D-6B9)

Description
Specificity: Recognises human CALR.
Specifications
Specifications
| Antigen | Calreticulin |
| Applications | ELISA, Flow Cytometry, Western Blot, Immunocytochemistry, Mass Spectrometry |
| Classification | Recombinant Monoclonal |
| Clone | SAIC-16D-6B9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | CALR |
| Gene Accession No. | P27797 |
| Gene Alias | Autoantigen RO; CABP3; CALBP; calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; calreticulin precursor; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99n; I79_008346; RO; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin); SSA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.