Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Calretinin Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1110R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS1110R1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS1110R1 Supplier Epredia™ Supplier No. MS1110R1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Antigen Calretinin Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5A5
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant full length mouse calretinin protein
Quantity 1 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.